Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tprg1
Entrez Gene ID
560596
Gene Name
tumor protein p63 regulated 1
Gene Aliases
zgc:162933
Chromosome Location
Chr.6: 98641 - 135987 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089528.2 NP_001082997.1 5 - 6 756 74
GenBank mRNA BC139672.1 - 5 - 6 757 74
BC158178.1 - 5 - 6 757 74

View Full Product Details