Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp4cb
Entrez Gene ID
562705
Gene Name
protein phosphatase 4, catalytic subunit b
Gene Aliases
PP4C-B, zgc:172126
Chromosome Location
Chr.12: 3964351 - 3978308 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111168.1 NP_001104638.1 1 - 2 106 141
GenBank mRNA BC155609.1 - 1 - 2 106 141

View Full Product Details