Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
olfm3a
Entrez Gene ID
563182
Gene Name
olfactomedin 3a
Gene Aliases
olfm3, zgc:171621
Chromosome Location
Chr.24: 28686305 - 28711176 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111172.1 NP_001104642.1 4 - 5 878 94
GenBank mRNA BC153993.1 - 4 - 5 878 94

View Full Product Details