Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
and2
Entrez Gene ID
566702
Gene Name
actinodin2
Gene Aliases
zgc:175106
Chromosome Location
Chr.2: 31763201 - 31767652 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111190.1 NP_001104660.1 4 - 5 326 136
GenBank mRNA BC155829.1 - 4 - 5 326 136

View Full Product Details