Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
il23r
Entrez Gene ID
100134972
Gene Name
interleukin 23 receptor
Gene Aliases
il-23r
Chromosome Location
Chr.6: 34869701 - 34895164 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113506.1 NP_001106978.1 1 - 2 217 65

View Full Product Details