Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ebi3
Entrez Gene ID
100134974
Gene Name
Epstein-Barr virus induced 3
Gene Aliases
il-27rb, si:dkey-97a13.4
Chromosome Location
Chr.2: 41936622 - 41942645 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113508.1 NP_001106980.1 1 - 2 138 60

View Full Product Details