Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hoxd4a
Entrez Gene ID
30329
Gene Name
homeobox D4a
Gene Aliases
etID309823.9, etID37000.9, fb36c05, hoxd4, wu:fb36c05
Chromosome Location
Chr.9: 1949366 - 1951004 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126445.2 NP_001119917.1 1 - 2 697 71
GenBank mRNA BC162966.1 - 1 - 2 697 71
BC162983.1 - 1 - 2 697 71
BC163059.1 - 1 - 2 693 71
Y14548.1 - - 837 71

View Full Product Details