Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hbae1.1
Entrez Gene ID
30597
Gene Name
hemoglobin, alpha embryonic 1.1
Gene Aliases
HBA2r, HBAA1, SI:dZ118J2.10, SI:dZ118J2.8, dZ118J2.6, hba2, hbae1, si:by119c3.1, si:by119c3.3, si:xx-119c3.3, wu:fa12a11, wu:fb74f06, zgc:86936
Chromosome Location
Chr.3: 55120372 - 55121066 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182940.3 NP_891985.1 2 - 3 341 55
GenBank mRNA AY325264.1 - 2 - 3 309 55
BC071550.1 - 2 - 3 341 55

Gene Symbol
hbae1.2
Entrez Gene ID
572729
Gene Name
hemoglobin, alpha embryonic 1.3
Gene Aliases
ae1b, hbae1b, si:ch211-5k11.2
Chromosome Location
Chr.3: 55127554 - 55128280 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001365350.1 NP_001352279.1 2 - 3 344 55
XM_001333519.7 XP_001333555.1 2 - 3 373 55

Gene Symbol
hbae1.3
Entrez Gene ID
100334599
Gene Name
hemoglobin, alpha embryonic 1.3
Gene Aliases
ae1c, hbae1c, si:ch211-5k11.2
Chromosome Location
Chr.3: 55138413 - 55139156 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001365351.1 NP_001352280.1 2 - 3 344 55
GenBank mRNA BC152154.1 - 2 - 3 341 55
BC155770.1 - 2 - 3 341 55

View Full Product Details