Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
phf23b
Entrez Gene ID
100004597
Gene Name
PHD finger protein 23b
Gene Aliases
wu:fi23e08, zgc:110484, zgc:66469
Chromosome Location
Chr.7: 26616242 - 26627253 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199844.1 NP_956138.1 5 - 6 1141 67
GenBank mRNA BC055673.1 - 5 - 6 1141 67
BC095292.1 - 4 - 5 1125 67
BC154223.1 - 4 - 5 1207 67
BC164060.1 - 4 - 5 789 67

View Full Product Details