Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ankmy2a
- Entrez Gene ID
-
334389
- Gene Name
- ankyrin repeat and MYND domain containing 2a
- Gene Aliases
-
fi46e08, wu:fi46e08, zgc:55491
- Chromosome Location
-
Chr.15: 34573311 - 34590820 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199894.1 |
NP_956188.1 |
1 - 2 |
154 |
100 |
GenBank mRNA |
BC044410.1 |
- |
1 - 2 |
154 |
100 |
- Gene Symbol
- ankmy2b
- Entrez Gene ID
-
100034633
- Gene Name
- ankyrin repeat and MYND domain containing 2b
- Gene Aliases
-
ankmy2, si:dkeyp-25a3.5
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CK698800.1 |
- |
1 - 2 |
159 |
100 |
View Full Product Details