Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrsn1
Entrez Gene ID
335551
Gene Name
neurensin 1
Gene Aliases
fj21c11, wu:fj21c11, zgc:73062
Chromosome Location
Chr.16: 217950 - 226654 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199959.1 NP_956253.1 2 - 3 243 96
GenBank mRNA BC060899.1 - 2 - 3 243 96

View Full Product Details