Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnase2
Entrez Gene ID
100000399
Gene Name
deoxyribonuclease II, lysosomal
Gene Aliases
fa56d08, fa92d01, wu:fa56d08, wu:fa92d01
Chromosome Location
Chr.16: 45337672 - 45357551 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114738.1 NP_001108210.1 3 - 4 342 122
XM_021466569.1 XP_021322244.1 3 - 4 392 122
GenBank mRNA BC159268.1 - 3 - 4 352 122

View Full Product Details