Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pcdh18a
Entrez Gene ID
572532
Gene Name
protocadherin 18a
Gene Aliases
id:ibd5114, pcdh18
Chromosome Location
Chr.1: 13261115 - 13271156 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115058.1 NP_001108530.1 1 - 2 2931 94
XM_005159847.4 XP_005159904.1 1 - 2 2985 94
GenBank mRNA EU267178.1 - 1 - 2 2931 94

View Full Product Details