Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
qtrt2
Entrez Gene ID
402798
Gene Name
queuine tRNA-ribosyltransferase accessory subunit 2
Gene Aliases
qtrtd1
Chromosome Location
Chr.24: 21059673 - 21090517 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001118896.2 NP_001112368.2 2 - 3 286 66
XM_005162680.4 XP_005162737.1 2 - 3 265 66
GenBank mRNA EH559864.1 - 5 - 6 595 66

View Full Product Details