Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ches1
Entrez Gene ID
555843
Gene Name
checkpoint suppressor 1
Gene Aliases
-
Chromosome Location
Chr.20: 16478141 - 16548903 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122745.1 NP_001116217.1 4 - 5 752 79
XM_005160419.4 XP_005160476.1 4 - 5 970 79

View Full Product Details