Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nfr
Entrez Gene ID
570084
Gene Name
notochord formation related
Gene Aliases
oor
Chromosome Location
Chr.5: 28497934 - 28503177 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_040606.1 2 - 3 111 104
GenBank mRNA EU520304.1 - 2 - 3 96 104

View Full Product Details