Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tnn
Entrez Gene ID
30234
Gene Name
tenascin N
Gene Aliases
tenw, tnw, zgc:110729
Chromosome Location
Chr.2: 35509418 - 35567016 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131036.2 NP_571111.2 10 - 11 2261 115
XM_005171264.4 XP_005171321.1 11 - 12 2520 115
XM_005171265.3 XP_005171322.1 9 - 10 1992 115
XM_009298633.3 XP_009296908.1 11 - 12 2256 115
GenBank mRNA AJ001423.1 - 10 - 11 2221 116
KM655642.1 - 9 - 10 2095 115

View Full Product Details