Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tnn
Entrez Gene ID
30234
Gene Name
tenascin N
Gene Aliases
tenw, tnw, zgc:110729
Chromosome Location
Chr.2: 35509418 - 35567016 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131036.2 NP_571111.2 7 - 8 1720 63
XM_005171264.4 XP_005171321.1 7 - 8 1715 63
XM_009298633.3 XP_009296908.1 7 - 8 1715 63
GenBank mRNA AJ001423.1 - 7 - 8 1680 63
KM655642.1 - 6 - 7 1554 63

View Full Product Details