Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
derl3
Entrez Gene ID
393877
Gene Name
derlin 3
Gene Aliases
zgc:63991
Chromosome Location
Chr.8: 154093 - 162333 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200903.2 NP_957197.2 4 - 5 536 92
GenBank mRNA BC064666.1 - 4 - 5 535 92
CK026866.1 - 4 - 5 462 92

View Full Product Details