Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttn.1
Entrez Gene ID
100001684
Gene Name
titin, tandem duplicate 1
Gene Aliases
TTN, hm:zeh1256r, im:6911926, im:7146708, si:dZ167C3.1, si:dz249n21.1, ttnb, ttnl, wu:fc09a12
Chromosome Location
Chr.9: 42732992 - 42873861 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021479071.1 XP_021334746.1 182 - 183 65897 74
XM_021479072.1 XP_021334747.1 140 - 141 58296 74
GenBank mRNA CK676433.1 - 1 - 2 161 74

View Full Product Details