Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aurkb
Entrez Gene ID
791894
Gene Name
aurora kinase B
Gene Aliases
CHUNP6896, aurB, cei, fa09g06, fd02h12, stka, wu:fa09g06, wu:fd02h12
Chromosome Location
Chr.14: 241461 - 246396 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212566.2 NP_997731.2 3 - 4 152 103
GenBank mRNA AI958300.1 - 2 - 3 141 103
AY099518.1 - 3 - 4 177 103
BC067695.1 - 3 - 4 160 103

View Full Product Details