Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttpal
Entrez Gene ID
321577
Gene Name
tocopherol (alpha) transfer protein-like
Gene Aliases
wu:fb20b02, wu:fj77b05, zgc:63845
Chromosome Location
Chr.23: 25868608 - 25874877 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212590.2 NP_997755.2 3 - 4 679 144
XM_021469732.1 XP_021325407.1 3 - 4 714 144
GenBank mRNA BC055264.1 - 3 - 4 668 144
EH434754.1 - 3 - 4 688 144

View Full Product Details