Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdzd11
Entrez Gene ID
405893
Gene Name
PDZ domain containing 11
Gene Aliases
zgc:77536
Chromosome Location
Chr.14: 228679 - 232473 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212957.1 NP_998122.1 1 - 2 124 74
GenBank mRNA BC067163.1 - 1 - 2 124 74
BC154539.1 - 2 - 3 136 74

View Full Product Details