Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pnp5a
Entrez Gene ID
791647
Gene Name
purine nucleoside phosphorylase 5a
Gene Aliases
np, wu:fd51h11, zgc:77008
Chromosome Location
Chr.7: 6652967 - 6668424 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213311.2 NP_998476.1 3 - 4 525 82
GenBank mRNA BC066610.1 - 3 - 4 464 82
EB931190.1 - 3 - 4 442 82

View Full Product Details