Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp1caa
Entrez Gene ID
407980
Gene Name
protein phosphatase 1, catalytic subunit, alpha isozyme a
Gene Aliases
wu:fc04c08, wu:fc09b07, wu:fc30g11, wu:fe05h08, zgc:85729
Chromosome Location
Chr.3: 480356 - 499413 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214811.2 NP_999976.1 6 - 7 1340 80
GenBank mRNA BC070008.1 - 6 - 7 1293 80
CD758806.1 - 4 - 5 690 80

View Full Product Details