Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPO3
Entrez Gene ID
100067377
Gene Name
R-spondin 3
Gene Aliases
-
Chromosome Location
Chr.10: 75066738 - 75143624 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109682.1 NP_001103152.1 1 - 2 98 76
GenBank mRNA EU029662.1 - 1 - 2 98 76

View Full Product Details