Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRISP3
Entrez Gene ID
100033965
Gene Name
cysteine-rich secretory protein 3
Gene Aliases
AEG2, HSP-3
Chromosome Location
Chr.20: 48736747 - 48761128 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081874.1 NP_001075343.1 3 - 4 243 67
XM_014734198.2 XP_014589684.2 - 295 67
GenBank mRNA AJ001400.1 - 3 - 4 243 67
AJ459964.1 - 3 - 4 243 67

View Full Product Details