Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGFS
Entrez Gene ID
100034026
Gene Name
prostaglandin F synthase
Gene Aliases
-
Chromosome Location
Chr.29: 30323578 - 30338871 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081895.2 NP_001075364.1 9 - 10 1117 146
XM_005606827.3 XP_005606884.1 - 1174 146
GenBank mRNA AY304536.1 - 9 - 10 1118 146

View Full Product Details