Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSD11B2
Entrez Gene ID
100034124
Gene Name
hydroxysteroid 11-beta dehydrogenase 2
Gene Aliases
11-HSD2
Chromosome Location
Chr.3: 18568978 - 18574777 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081926.2 NP_001075395.1 4 - 5 799 69
GenBank mRNA AF126744.1 - 4 - 5 799 69
AF126745.1 - 4 - 5 799 69

View Full Product Details