Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIBIN
Entrez Gene ID
100057578
Gene Name
fin bud initiation factor homolog
Gene Aliases
-
Chromosome Location
Chr.7: 95777120 - 95778745 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099443.1 NP_001092913.1 1 - 1 1448 131
GenBank mRNA AB302195.1 - 1 - 1 1448 131

View Full Product Details