Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS4
Entrez Gene ID
100033914
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 4
Gene Aliases
-
Chromosome Location
Chr.5: 32402597 - 32409605 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111299.2 NP_001104769.2 4 - 5 1260 90
GenBank mRNA AF368321.1 - 2 - 3 267 90
EU025848.1 - 4 - 5 1260 90

View Full Product Details