Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFNB3
Entrez Gene ID
100062080
Gene Name
ephrin B3
Gene Aliases
-
Chromosome Location
Chr.11: 50981283 - 50985237 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114942.1 NP_001108414.1 1 - 2 126 71
GenBank mRNA EF682146.1 - 1 - 2 126 71

View Full Product Details