Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL17A
Entrez Gene ID
100034142
Gene Name
interleukin 17A
Gene Aliases
IL17
Chromosome Location
Chr.20: 50879963 - 50883546 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143792.1 NP_001137264.1 2 - 3 225 95
XM_023624010.1 XP_023479778.1 - 355 95
GenBank mRNA AY014959.1 - 1 - 2 169 95
EU744563.2 - 2 - 3 225 95
KX345375.1 - 1 - 2 156 95

View Full Product Details