Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FASLG
Entrez Gene ID
100052326
Gene Name
Fas ligand
Gene Aliases
TNLG1A, fasL
Chromosome Location
Chr.5: 8587268 - 8594020 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166039.2 NP_001159511.1 2 - 3 401 86
GenBank mRNA GQ429291.1 - 2 - 3 377 86

View Full Product Details