Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPC3
Entrez Gene ID
100302031
Gene Name
two pore channel 3
Gene Aliases
-
Chromosome Location
Chr.15: 14160976 - 14199717 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001245905.1 NP_001232834.1 2 - 3 225 107
XM_023618423.1 XP_023474191.1 - 319 107

View Full Product Details