Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAMLD1
Entrez Gene ID
100069293
Gene Name
mastermind like domain containing 1
Gene Aliases
f18
Chromosome Location
Chr.X: 123396878 - 123521456 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173458.1 NP_001166929.1 1 - 2 95 67
XM_005614568.3 XP_005614625.2 - 340 67
XM_005614569.3 XP_005614626.2 - 340 67
GenBank mRNA GQ869480.1 - 1 - 2 95 67

View Full Product Details