Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAA1
Entrez Gene ID
100034017
Gene Name
serum amyloid A1
Gene Aliases
SAA
Chromosome Location
Chr.7: 88539140 - 88543005 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163892.1 NP_001157364.1 4 - 4 513 84

View Full Product Details