Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCAIM
Entrez Gene ID
420670
Gene Name
T-cell activation inhibitor, mitochondrial
Gene Aliases
C2H3ORF23, C3orf23, TOAG-1, TOAG1
Chromosome Location
Chr.2: 40558061 - 40581115 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286225.1 NP_001273154.1 1 - 2 108 101
XM_015281494.2 XP_015136980.1 2 - 3 195 101
XM_015281495.2 XP_015136981.1 2 - 3 1090 101
XM_015281496.2 XP_015136982.1 2 - 3 131 101
XM_015281497.2 XP_015136983.1 2 - 3 435 101
XM_025147211.1 XP_025002979.1 2 - 3 183 101
XM_025147212.1 XP_025002980.1 1 - 2 1605 101
GenBank mRNA BX930726.1 - 2 - 3 182 101

View Full Product Details