Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NABP1
Entrez Gene ID
424046
Gene Name
nucleic acid binding protein 1
Gene Aliases
OBFC2A
Chromosome Location
Chr.7: 8141843 - 8157061 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_421902.6 XP_421902.2 2 - 3 457 92
GenBank mRNA BX931186.1 - 2 - 3 439 92

View Full Product Details