Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CETN1
Entrez Gene ID
422303
Gene Name
centrin 1
Gene Aliases
CETN2
Chromosome Location
Chr.4: 11283946 - 11288350 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277640.1 NP_001264569.1 4 - 5 481 73
GenBank mRNA BU113047.1 - 4 - 5 479 73
BU115307.1 - 4 - 5 480 73
BX933907.1 - 4 - 5 481 73

View Full Product Details