Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAPB
Entrez Gene ID
421231
Gene Name
NSF attachment protein beta
Gene Aliases
-
Chromosome Location
Chr.3: 3276268 - 3283076 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199430.1 NP_001186359.1 10 - 11 847 73
GenBank mRNA BX933953.1 - 9 - 10 670 73
CR386813.1 - 4 - 5 326 73

View Full Product Details