Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC15
Entrez Gene ID
418838
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Gene Aliases
DNAJD1
Chromosome Location
Chr.1: 168054028 - 168078478 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277776.1 NP_001264705.1 3 - 4 438 90
NM_001277777.1 NP_001264706.1 3 - 4 167 90
GenBank mRNA BU363480.1 - 3 - 4 312 90
BX929634.2 - 3 - 4 312 90
BX933235.1 - 3 - 4 295 90
BX935210.2 - 3 - 4 438 90
CN234998.1 - 3 - 4 170 90

View Full Product Details