Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX13
Entrez Gene ID
421197
Gene Name
peroxisomal biogenesis factor 13
Gene Aliases
-
Chromosome Location
Chr.3: 2124834 - 2132004 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199421.1 NP_001186350.1 2 - 3 879 82
GenBank mRNA BX930866.2 - 1 - 2 87 82
BX935703.1 - 2 - 3 773 82

View Full Product Details