Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOX3
Entrez Gene ID
415724
Gene Name
TOX high mobility group box family member 3
Gene Aliases
TNRC9
Chromosome Location
Chr.11: 5223979 - 5345307 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX933453.2 - 3 - 4 446 153

View Full Product Details