Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACYP2
Entrez Gene ID
421217
Gene Name
acylphosphatase 2
Gene Aliases
acylphosphatase-2
Chromosome Location
Chr.3: 2882010 - 2928755 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167762.1 NP_001161234.1 1 - 2 144 70
XM_025148627.1 XP_025004395.1 1 - 2 200 70
GenBank mRNA BG710739.1 - 1 - 2 119 70
BU393960.1 - 1 - 2 144 70
CR353528.1 - 1 - 2 142 70

View Full Product Details