Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LSM11
Entrez Gene ID
416241
Gene Name
LSM11, U7 small nuclear RNA associated
Gene Aliases
-
Chromosome Location
Chr.13: 11997558 - 12008663 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX950474.2 - 4 - 5 725 84

View Full Product Details