Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUSP12
Entrez Gene ID
425349
Gene Name
dual specificity phosphatase 12
Gene Aliases
-
Chromosome Location
Chr.1: 79964646 - 79968560 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278099.3 NP_001265028.1 2 - 3 394 57
GenBank mRNA BU428045.1 - 2 - 3 214 57
CN219687.1 - 2 - 3 394 57
CR354240.1 - 2 - 3 229 57
CR387446.1 - 2 - 3 388 57

View Full Product Details