Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNAP47
Entrez Gene ID
420399
Gene Name
synaptosomal associated protein 47
Gene Aliases
C1orf142
Chromosome Location
Chr.2: 3277464 - 3303728 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004939091.3 XP_004939148.1 5 - 5 3049 73
XM_004939093.3 XP_004939150.1 5 - 5 4104 73
XM_418505.6 XP_418505.4 5 - 5 2879 73
GenBank mRNA CR389677.1 - 1 - 1 1545 73

View Full Product Details