Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIPRL
Entrez Gene ID
418374
Gene Name
TOR signaling pathway regulator
Gene Aliases
-
Chromosome Location
Chr.1: 85239455 - 85248961 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_416593.6 XP_416593.1 4 - 5 668 89
GenBank mRNA BX935082.2 - 4 - 5 638 89
BX950267.2 - 4 - 5 616 89
CR406044.1 - 4 - 5 634 89

View Full Product Details