Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UNC80
Entrez Gene ID
424003
Gene Name
unc-80 homolog, NALCN activator
Gene Aliases
C2ORF21
Chromosome Location
Chr.7: 2587617 - 2709204 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025152713.1 XP_025008481.1 19 - 20 3357 76
XM_025152718.1 XP_025008486.1 19 - 20 3241 76
GenBank mRNA CR406629.1 - 2 - 3 298 76

View Full Product Details